missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) nitride, 99.99% (metals basis)

Catalog No. AA4021818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4021818 50 g
AA4021809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4021818 Supplier Thermo Scientific Chemicals Supplier No. 04021818

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25617-97-4 | GaN | 83.73 g/mol

Gallium(III) nitride is used to read blu-ray discs. It is used in various wireless infrastructure applications. It finds applications in optoelectronic, high-power and high-frequency devices. It is also useful in military electronics (active electronically scanned array radars). It is biocompatible and used in electrodes and electronic components in living organisms. It is a suitable material for solar cell arrays for satellites. It is employed in bright light-emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25617-97-4
Molecular Formula GaN
Molecular Weight (g/mol) 83.73
MDL Number MFCD00016108
InChI Key JMASRVWKEDWRBT-UHFFFAOYSA-N
Synonym gallium nitride, gallium mononitride, unii-1r9cc3p9vl, gallium nitride,, 1r9cc3p9vl, gallium nitride trace metals basis, gallium iii nitride trace metals basis 10g
PubChem CID 117559
IUPAC Name azanylidynegallane
SMILES N#[Ga]

Specifications

Melting Point 800°C (sublimation)
Color Yellow
Physical Form Powder
Quantity 50 g
Sensitivity Moisture sensitive
Density 6.1 g/mL
Merck Index 14,4351
Solubility Information Slightly soluble in hot concentrated sulfuric acid and hot conc; sodium hydroxide; Insoluble in water and dilute acids.
Formula Weight 83.73
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Gallium(III) nitride
Show More Show Less
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 247-129-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only