missing translation for 'onlineSavingsMsg'
Learn More

Gallic Acid, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111013192
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013192 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013192 Supplier Merck Emd Millipore Supplier No. 91215100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 149-91-7
Product Line TraceCERT
Physical Form Neat
Linear Formula (HO)3C6H2CO2H
MDL Number MFCD00002510
Melting Point 251°C (decomposition)
Synonym 3,4,5-Trihydroxybenzoic acid
Quantity 100 mg
Formula Weight 170.12
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC