missing translation for 'onlineSavingsMsg'
Learn More

Galena, naturally occurring mineral, approximately 1-2in

Catalog No. AA42546KJ
Change view
Click to view available options
Quantity:
1 Ea.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42546KJ 5 Pc.
AA42546KF 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42546KJ Supplier Thermo Scientific Chemicals Supplier No. 042546KJ

CAS: 1314-87-0 | PbS | 239.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
PubChem CID 14819
IUPAC Name sulfanylidenelead
SMILES [S--].[Pb++]

Specifications

Physical Form Crystal Clusters
Quantity 5 Pc.
UN Number UN3077
Solubility Information Soluble in strong HNO3,in excess of hot HCl
Formula Weight 239.25
Packaging Bag
Chemical Name or Material Galena
Hazard Category H302+H332-H350-H360Df-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 215-246-6
RTECSNumber OG4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only