Learn More
Eisco™ Galena Mineral Specimen
Catalog No. S24688
Change view
Click to view available options
Quantity:
12/Pk.
Catalog No. Quantity
S24688 12/Pk.
1 options
Catalog No. S24688 Supplier Eisco™ Supplier No. ESNG0015PK12
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use as tool to explain specific gravity to students - EISCO Galena Mineral Specimen is used in classroom identification labs and exercises, correlates well to Next Generation Science Standards Evidence Statements.
  • Far denser than most minerals
  • Reflective cubic crystals offer shining example of metallic luster
  • Comes in clear labeled and barcoded bag for storage
Specifications
Quantity 12/Pk.
For Use With (Application) Study of specific gravity
Product Type Galena Mineral Study Specimen
Width (English) Approx. 1 in.
Width (Metric) Approx. 3 cm
Packaging Clear labeled and barcoded bag