missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galectin 2 Monoclonal Antibody (C6)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA543814

 Afficher plus de versions de ce produit

Numéro de catalogue. PIMA543814


Il en reste null

Description

Description

Galectin 2 Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction.
Spécifications

Identifiants chimiques

Galectin 2
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Western Blot
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P05162, Q29373, Q9CQW5, Q9Z144
100217393, 107753, 171134, 3957
Recombinant protein Galectin 2. The antigen corresponds to amino acid range 1-132 of the target protein.
Antibody
C6
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG2b κ
Human, Mouse, Rat, Pig
LGALS2
2200008F12Rik; AI324147; Beta-galactoside-binding lectin L-14-II; gal-2; galectin 2; galectin-2; HL14; lactose-binding lectin 2; lectin galactoside-binding soluble 2 (galectin 2); lectin, galactose-binding, soluble 2; lectin, galactoside binding soluble 2; lectin, galactoside-binding, soluble, 2; lectin, galactoside-binding, soluble, 2 (galectin 2); LEG2; Lgals2; S-Lac lectin 2
LGALS2
Primary
Protein A/G

Spécifications

Galectin 2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P05162, Q29373, Q9CQW5, Q9Z144
LGALS2
Recombinant protein Galectin 2. The antigen corresponds to amino acid range 1-132 of the target protein.
20 μL
Primary
Human, Mouse, Rat, Pig
Antibody
IgG2b κ
Immunohistochemistry (Paraffin), Western Blot, Western Blot
C6
Unconjugated
LGALS2
2200008F12Rik; AI324147; Beta-galactoside-binding lectin L-14-II; gal-2; galectin 2; galectin-2; HL14; lactose-binding lectin 2; lectin galactoside-binding soluble 2 (galectin 2); lectin, galactose-binding, soluble 2; lectin, galactoside binding soluble 2; lectin, galactoside-binding, soluble, 2; lectin, galactoside-binding, soluble, 2 (galectin 2); LEG2; Lgals2; S-Lac lectin 2
Mouse
Protein A/G
RUO
100217393, 107753, 171134, 3957
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions