missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 10 Monoclonal Antibody (C2)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566040
Description
Galectin 10 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Regulates immune responses through the recognition of cell-surface glycans. Essential for the anergy and suppressive function of CD25-positive regulatory T-cells (Treg).Identifiants chimiques
| Galectin 10 | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q05315 | |
| 1178 | |
| Recombinant Charcot Leyden Crystal Protein (CLC) (Ser2-Arg142). | |
| Antibody |
| C2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human | |
| CLC | |
| Charcot-Leyden crystal galectin; Charcot-Leyden crystal protein; CLC; EC 3.1.1.5; Eosinophil lysophospholipase; GAL10; Gal-10; galectin 10; galectin-10; lectin, galactoside-binding, soluble, 10; LGALS10; LGALS10A; LPPL_HUMAN; lysolecithin acylhydrolase; MGC149659 | |
| CLC | |
| Primary | |
| Protein A/G |
Spécifications
| Galectin 10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q05315 | |
| CLC | |
| Recombinant Charcot Leyden Crystal Protein (CLC) (Ser2-Arg142). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C2 | |
| Unconjugated | |
| CLC | |
| Charcot-Leyden crystal galectin; Charcot-Leyden crystal protein; CLC; EC 3.1.1.5; Eosinophil lysophospholipase; GAL10; Gal-10; galectin 10; galectin-10; lectin, galactoside-binding, soluble, 10; LGALS10; LGALS10A; LPPL_HUMAN; lysolecithin acylhydrolase; MGC149659 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1178 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |