missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)

Catalog No. AC316760250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC316760250 25 g Glass bottle
AC316761000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC316760250 Supplier Thermo Scientific Chemicals Supplier No. 316760250

CAS: 19598-90-4 | GdN3O9·6H2O | 451.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 19598-90-4
Molecular Formula GdN3O9·6H2O
Molecular Weight (g/mol) 451.4
MDL Number MFCD00149728
InChI Key XWFVFZQEDMDSET-UHFFFAOYSA-N
Synonym gadolinium iii nitrate hexahydrate, gadolinium nitrate hexahydrate, gadolinium iii nitrate, hexahydrate 1:3:6, nitric acid, gadolinium 3+ salt, hexahydrate, gadoliniumnitratehexahydrate, gd.3no3.6h2o, gadolinium 3+ trinitrate hexahydrate, gadolinium nitrate, hexahydrate,, nitric acid,gadolinium 3+ salt, hexahydrate 8ci,9ci, gadolinium iii nitrate hexahydrate trace metals basis
PubChem CID 209258
IUPAC Name gadolinium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Gd+3]

Specifications

Melting Point 91°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99.90%
Linear Formula Gd(NO3)3·6H2O
Solubility Information Solubility in water: soluble
Formula Weight 451.4
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) nitrate hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

RUO – Research Use Only