missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium ICP Standard, 1000 ppm Gd in 3% HNO3, Ricca Chemical

Catalog No. p-3251705
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PGD1KN100 Natural LDPE Bottle
PGD1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PGD1KN100 Supplier Ricca Chemical Company Supplier No. PGD1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12064-62-9 , 7697-37-2 , 7732-18-5
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.497
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
PubChem CID 159427
IUPAC Name gadolinium(3+);oxygen(2-)
SMILES [O-2].[O-2].[O-2].[Gd+3].[Gd+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Gd
CAS Min % 93.0, 4.56, 0.11
CAS Max % 97.0, 4.74, 0.11
Equivalence 1 mL = 1 mg Gd
Chemical Name or Material Gadolinium ICP Standard