Learn More
G-Biosciences G-Trap™ Immobilized D-Alanine Column
Description
Immobilized Amino Acids are prepared by coupling amino acid to preactivated 6% cross-linked agarose. The covalent coupling to agarose (resin) is achieved by cross-linking the amino or sulfhydryl group of the amino acid to the preactivated resin.
Spécifications
Spécifications
| Product Line | GeneBLAzer |
| Product Type | Immobilized D-Alanine |
| Quantity | 5 mL |