missing translation for 'onlineSavingsMsg'
Learn More

Fusidic Acid 97.0+%, TCI America™

Catalog No. F10075G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10075G Supplier TCI America Supplier No. F10075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6990-06-3
Molecular Formula C31H48O6
Molecular Weight (g/mol) 516.72
MDL Number MFCD00865135
InChI Key IECPWNUMDGFDKC-MZJAQBGESA-N
Synonym fusidic acid, fusidine, ramycin, fucithalmic, fucidic acid, fucidin acid, fusidate, fucidin, flucidin, fucidate
PubChem CID 3000226
ChEBI CHEBI:29013
IUPAC Name (2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid
SMILES CC1C2CCC3(C(C2(CCC1O)C)C(CC4C3(CC(C4=C(CCC=C(C)C)C(=O)O)OC(=O)C)C)O)C

Specifications

Melting Point 193°C
Color White
Quantity 5 g
Formula Weight 516.72
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fusidic Acid
RTECSNumber RC1350000
TSCA No
Recommended Storage Refrigerator