missing translation for 'onlineSavingsMsg'
Learn More

Fusaric acid, For HPLC derivatization (HPLC), ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014514
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014514 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014514 Supplier Merck Emd Millipore Supplier No. 559521G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00006298
Synonym 5-Butylpicolinic acid; 5-Butylpyridine-2-carboxylic acid

Specifications

CAS 536-69-6
Linear Formula C10H13 NO2
Quantity 1 g
Formula Weight 179.22
Percent Purity ≥99.0% (HPLC)
Grade For HPLC derivatization
RTECSNumber US5625000