missing translation for 'onlineSavingsMsg'
Learn More

Furosemide 99.0+%, TCI America™

Supplier:  TCI America F018225G

 View more versions of this product

Catalog No. F018225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Furosemide
205°C
C12H11ClN2O5S
25 g
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
3440
330.74
Crystalline Powder
54-31-9
White
MFCD00010549
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl
330.739
CHEBI:47426
≥99.0% (T)

Chemical Identifiers

54-31-9
330.739
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
3440
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
C12H11ClN2O5S
MFCD00010549
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
CHEBI:47426
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CB2625000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions