accessibility menu, dialog, popup

UserName

CAS RN 54-31-9

IUPAC Name: 4-chloro-2-{[(furan-2-yl)methyl]amino}-5-sulfamoylbenzoic acid
Synonyms: furosemide dihydroflumethiazide methforylthiazidine transit frusemid fursemid metflorylthiazidine salix furosemid furosemidu
Molecular Weight (g/mol): 330.74
Molecular Formula: C12H11ClN2O5S
InChi Key: ZZUFCTLCJUWOSV-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC(C(O)=O)=C(NCC2=CC=CO2)C=C1Cl
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)