missing translation for 'onlineSavingsMsg'
Learn More

Furosemide 99.0+%, TCI America™

Catalog No. p-7129523
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
F01825G 5 g
F018225G 25 g
2 options

Catalog No. F01825G

Supplier: TCI America F01825G

Only null left

Chemical Identifiers

CAS 54-31-9
Molecular Formula C12H11ClN2O5S
Molecular Weight (g/mol) 330.739
MDL Number MFCD00010549
InChI Key ZZUFCTLCJUWOSV-UHFFFAOYSA-N
Synonym furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
PubChem CID 3440
ChEBI CHEBI:47426
IUPAC Name 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
SMILES C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl

Specifications

Melting Point 205°C
Color White
Quantity 5 g
Formula Weight 330.74
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Furosemide
RTECSNumber CB2625000
TSCA No