missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Mercaptan 98.0+%, TCI America™

Catalog No. F0077500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0077500G 500 g
F007725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0077500G Supplier TCI America Supplier No. F0077500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-02-2
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.162
MDL Number MFCD00003254
InChI Key ZFFTZDQKIXPDAF-UHFFFAOYSA-N
Synonym furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol
PubChem CID 7363
IUPAC Name furan-2-ylmethanethiol
SMILES C1=COC(=C1)CS

Specifications

Color Yellow
Boiling Point 155°C
Quantity 500 g
UN Number 1228
Formula Weight 114.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfuryl Mercaptan
EINECSNumber (5)-0050
RTECSNumber LU2100000
TSCA Yes