missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Alcohol 98.0+%, TCI America™

Catalog No. F007625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F007625ML 25 mL
F0076500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F007625ML Supplier TCI America Supplier No. F007625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-00-0
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00003252
InChI Key XPFVYQJUAUNWIW-UHFFFAOYSA-N
Synonym furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
PubChem CID 7361
ChEBI CHEBI:207496
IUPAC Name furan-2-ylmethanol
SMILES C1=COC(=C1)CO

Specifications

Melting Point -29°C
Color Yellow
Boiling Point 170°C
Quantity 25 mL
UN Number 2874
Formula Weight 98.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfuryl Alcohol
EINECSNumber (5)-0031
RTECSNumber LU9100000
TSCA Yes