missing translation for 'onlineSavingsMsg'
Learn More

Furfural, Analytical Standard, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019368
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019368 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019368 Supplier Merck Emd Millipore Supplier No. 046231ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Furfural is a furan derivative obtained from renewable biomass-derived carbohydrates.

Specifications

CAS 98-01-1
Vapor Pressure 13.5 mmHg (55°C)
Vapor Density 3.31 (vs. air)
Density 1.16 g/mL (at 25°C)
Linear Formula C5H4O2
MDL Number MFCD00003229
Melting Point -36°C
Boiling Point 162°C
Synonym 2-Furaldehyde; Furan-2-carboxaldehyde
Quantity 1 mL
Formula Weight 96.08
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less