missing translation for 'onlineSavingsMsg'
Learn More

Furazolidone 98.0+%, TCI America™

Catalog No. F082125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F082125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F082125G Supplier TCI America Supplier No. F082125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-45-8
Molecular Formula C8H7N3O5
Molecular Weight (g/mol) 225.16
MDL Number MFCD00910550
InChI Key PLHJDBGFXBMTGZ-WEVVVXLNSA-N
Synonym furazolidone, furoxone, nitrofurazolidone, furazolidine, nitrofuroxon, furazolidon, diafuron, furaxone, furazol, furazon
PubChem CID 5323714
IUPAC Name 3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
SMILES C1COC(=O)N1N=CC2=CC=C(O2)[N+](=O)[O-]

Specifications

Melting Point 257°C
Color Yellow
Quantity 25 g
Formula Weight 225.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Furazolidone
EINECSNumber (9)-0991
RTECSNumber RQ3675000
TSCA Yes