missing translation for 'onlineSavingsMsg'
Learn More

Furan (stabilized with BHT) 99.0+%, TCI America™

Catalog No. F0074500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0074500ML 500 mL
F007425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0074500ML Supplier TCI America Supplier No. F0074500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-00-9
Molecular Formula C4H4O
Molecular Weight (g/mol) 68.08
MDL Number MFCD00003222
InChI Key YLQBMQCUIZJEEH-UHFFFAOYSA-N
Synonym divinylene oxide, furfuran, tetrole, oxole, oxacyclopentadiene, furane, 1,4-epoxy-1,3-butadiene, axole, rcra waste number u124, furfurane
PubChem CID 8029
ChEBI CHEBI:35559
IUPAC Name furan
SMILES O1C=CC=C1

Specifications

Melting Point -84°C
Color Yellow
Boiling Point 31°C
Quantity 500 mL
UN Number 2389
Formula Weight 68.08
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Furan (stabilized with BHT)
EINECSNumber (5)-3334
RTECSNumber LT8524000
TSCA Yes