missing translation for 'onlineSavingsMsg'
Learn More

Furan, 99%, stab. with ca 250ppm BHT

Catalog No. AAA13102AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13102AP 500 mL
AAA13102AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA13102AP Supplier Thermo Scientific Chemicals Supplier No. A13102AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-00-9 | C4H4O | 68.08 g/mol

Furan has an important biological properties and pharmacological activity. Its iodinated lipophilic derivative is widely used in the treatment of ventricular and arterial fibrillation. Its derivatives are used as inhibitors of biofilm formation for several bacterial species, as well as possess quorum-sensing inhibitory activities. 2-methylfuran and 2,5-dimethylfuran are used as lignocellulosic biofuels. Its resin is used to stop sand production in underground gas storage wells. It undergo Diels-Alder reaction with arynes to give derivatives of dihydronaphthalenes which are useful intermediates in synthesis of other polycyclic aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furan has an important biological properties and pharmacological activity. Its iodinated lipophilic derivative is widely used in the treatment of ventricular and arterial fibrillation. Its derivatives are used as inhibitors of biofilm formation for several bacterial species, as well as possess quorum-sensing inhibitory activities. 2-methylfuran and 2,5-dimethylfuran are used as lignocellulosic biofuels. Its resin is used to stop sand production in underground gas storage wells. It undergo Diels-Alder reaction with arynes to give derivatives of dihydronaphthalenes which are useful intermediates in synthesis of other polycyclic aromatic compounds.

Solubility
Slightly soluble in water. Soluble in common organic solvents, including alcohol, ether and acetone.

Notes
Light, heat and air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 110-00-9
Molecular Formula C4H4O
Molecular Weight (g/mol) 68.08
MDL Number MFCD00003222
InChI Key YLQBMQCUIZJEEH-UHFFFAOYSA-N
Synonym divinylene oxide, furfuran, tetrole, oxole, oxacyclopentadiene, furane, 1,4-epoxy-1,3-butadiene, axole, rcra waste number u124, furfurane
PubChem CID 8029
ChEBI CHEBI:35559
IUPAC Name furan
SMILES O1C=CC=C1

Specifications

Melting Point -86°C
Density 0.936
Boiling Point 32°C to 33°C
Flash Point -35°C (-31°F)
Refractive Index 1.422
Quantity 500 mL
UN Number UN2389
Beilstein 103221
Sensitivity Air and light sensitive
Merck Index 14,4296
Solubility Information Slightly soluble in water. Soluble in common organic solvents,including alcohol,ether and acetone.
Formula Weight 68.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Furan, stabilized with ≈250ppm BHT
Show More Show Less
Hazard Category H224-H302+H332-H315-H319-H341-H350-H373
Hazard Statement GHS H Statement

H224-H350-H341-H373-H302-H332-H315

Extremely flammable liquid and vapour.

May cause cancer.

Suspected of causing genetic defects.

May cause damage to organs through prolonged or repeated exposure.

Harmful if swallowed.

Harmful if inhaled.

Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FURAN
EINECSNumber 203-727-3
RTECSNumber LT8524000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only