missing translation for 'onlineSavingsMsg'
Learn More

Mopec Fume Exhaust Hood

Catalog No. 22444006
Change view
Click to view available options
Height (Metric):
50.8 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
22444006 50.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22444006 Supplier Mopec Supplier No. BF724

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Strong and lightweight solution for extracting fumes from the encapsulated work area. Mopec™ Fume Exhaust Hood is designed to isolate and remove potentially hazardous organic vapors from the laboratory environment.

  • Extruded aluminum frame construction and high impact LexanTM
  • Require one exhaust module

Warranty

1 Year Warranty

Specifications

Product Type Exhaust Hood
Description Requires ones exhaust module
Depth (English) 20 in.
Depth (Metric) 50.8 cm
Height (English) 20 in.
Width (English) 24 in.
Height (Metric) 50.8 cm
Width (Metric) 60.96cm