missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid, FCC, 99.5-100.5%, Spectrum™ Chemical

FU105, 110-17-8, C4H4O4

$639.09 - $6,607.18

Chemical Identifiers

CAS 110-17-8
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00002700
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18602866
View Documents Promotion Details
N/A 500 g Poly Bottle
Each for $639.09
Only null left
 
18602867
View Documents Promotion Details
N/A 2.5 kg Poly Bottle
Each for $1,860.33
Only null left
 
18602868
View Documents Promotion Details
N/A 12 kg Poly Bottle
Each for $3,459.79
Only null left
 
18602869
View Documents Promotion Details
N/A 50 kg Poly Bottle
Each for $6,607.18
Only null left
 
Description

Description

Spectrum™ Chemical Fumaric Acid, FCC is used as an acidity regulator and coagulant food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

110-17-8
116.07
VZCYOOQTPOCHFL-OWOJBTEDSA-N
OC(=O)\C=C\C(O)=O
C4H4O4
MFCD00002700
(2E)-but-2-enedioic acid

Specifications

110-17-8
0.1%
MFCD00002700
OC(=O)\C=C\C(O)=O
116.07
FCC
Fumaric Acid
100%
C4H4O4
VZCYOOQTPOCHFL-OWOJBTEDSA-N
(2E)-but-2-enedioic acid
99.5 to 100.5%
Crystalline Powder or Lumps
Videos
SDS
Documents

Documents