missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid, FCC, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18602866
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602866 500 g Poly Bottle
18602867 2.5 kg Poly Bottle
18602868 12 kg Poly Bottle
18602869 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602866 Supplier Spectrum Chemical Mfg Cor Supplier No. FU105500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

FU105, 110-17-8, C4H4O4

Spectrum™ Chemical Fumaric Acid, FCC is used as an acidity regulator and coagulant food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 110-17-8
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00002700
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.5 to 100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Fumaric Acid