accessibility menu, dialog, popup

UserName

OHOOHO(E)

CAS RN 110-17-8

IUPAC Name: (2E)-but-2-enedioic acid
Synonyms: fumaric acid monosodium salt maleic acid fumaric acid (substance) tumaric acid fumarate dianion disodium salt maleic acid scotchbond multipurpose etchant fema number 2488 dipotassium salt maleic acid iron salt maleic acid
Molecular Weight (g/mol): 116.07
Molecular Formula: C4H4O4
InChi Key: VZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILES: OC(=O)\C=C\C(O)=O
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (9)
  • (2)
Grade 
  • (4)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (15)
  • (9)
  • (2)
  • (9)
  • (2)
  • (7)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (9)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (9)
  • (2)
  • (4)
  • (5)
  • (1)
  • (5)