missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaric Acid, FCC, 99.5-100.5%, Spectrum™ Chemical
FU105, 110-17-8, C4H4O4
Supplier: Spectrum Chemical Mfg Cor FU1052.5KG
| Quantity | 2.5 kg |
|---|---|
| Packaging | Poly Bottle |
Description
Spectrum™ Chemical Fumaric Acid, FCC is used as an acidity regulator and coagulant food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 110-17-8 | |
| 116.07 | |
| VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
| OC(=O)\C=C\C(O)=O |
| C4H4O4 | |
| MFCD00002700 | |
| (2E)-but-2-enedioic acid |
Specifications
| Fumaric Acid | |
| 100% | |
| Poly Bottle | |
| 2.5 kg | |
| VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
| (2E)-but-2-enedioic acid | |
| 99.5 to 100.5% | |
| Crystalline Powder or Lumps |
| 110-17-8 | |
| 0.1% | |
| C4H4O4 | |
| MFCD00002700 | |
| OC(=O)\C=C\C(O)=O | |
| 116.07 | |
| FCC |