Learn More
Bioassay Systems FUMARATE ASSAY KIT, 100 TESTS DFS Item
Supplier:  Bioassay Systems EFUM100
Metabolism. EnzyChrom Fumarate Assay Kit. Method: OD565nm; Samples: Food, beverage and other biological samples (e.g. cell lysate, tissue homogenate, serum); Species: All; Procedure: 30 min; Size: 100 tests; Detection Limit: 5 uM; Shelf Life: 6 months
Catalog No. 501078336
Only null left
Explore available promotions