missing translation for 'onlineSavingsMsg'
Learn More

Fucoidan, Thermo Scientific™

Catalog No. AC467460010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467460010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC467460010 Supplier Thermo Scientific Chemicals Supplier No. 467460010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9072-19-9
MDL Number 00131109

Specifications

Alpha Vector FUCOIDAN
Color Light yellow to brown
Loss on Drying ≤10 %
Packaging Glass Bottle
Quantity 1 g
Physical Form Powder
Chemical Name or Material Fucoidan
Recommended Storage Normal conditions