missing translation for 'onlineSavingsMsg'
Learn More

FSI Triage Tape Systems

Catalog No. 19091018
Encompass
Change view
Click to view available options
Color:
Red, Yellow, Green, Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19091018 Red, Yellow, Green, Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19091018 Supplier FSI Supplier No. FSIEM19

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choose from tapes for standard triage or hazardous materials decontamination triage

Preassembled, portable kits dispense vinyl flagging tape for use during field triage. Tapes are transported in a cylindrical, Cordura™ nylon case that can be worn over the shoulder or as a fanny pack. Both cases feature Velcro™ closures, an adjustable web belt/strap, and a START initial triage algorithm quick reference protected by a clear plastic sleeve. Individual rolls measure approximately 1 in.W x 300 ft.L (2.5cm x 91.4m).

Triage-Plus Tape System

  • Contains one roll each of red, yellow, green, and black
  • Case measures 5 dia. x 8 in.L (13 x 20cm)

Triage/Decon Tape System

  • Contains one roll each of orange, blue, red, yellow, and black—orange tape identifies responsive contaminated patients; blue tape identifies unresponsive contaminated patients
  • Case measures 5 dia. x 10 in.L (13 x 25cm)

Specifications

Product Type Triage Plus Tape System
Color Red, Yellow, Green, Black
Dimensions (L x W) 300 ft. x 1 in. (91.4 m x 2.5 cm)