Learn More
Vileda Professional™ Roll-O-Matic™ Self-Wringing Mops
Suitable for all controlled environments.
Supplier: Vileda Professional™ FBG126104
Description
- Mop with built-in wringer
- Wringer built into handle, eliminating need for expensive and bulky wringer
- Improved components eliminate sharp edges and provide for excellent refill retention
- Fits all original and advanced technology Roll-O-Matic approved refills
- Ergonomic grip
Chemical Identifiers
| Blue | |
| For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique |
Specifications
| 72 x 14 in. | |
| Blue | |
| 14 in. | |
| Stainless Steel |
| Mop Handle | |
| 72 in. | |
| For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique |