missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Free Lime Reagent, Reagents
Free Lime Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$4389.11 - $9997.32
Chemical Identifiers
| CAS | 78-83-1 |
|---|---|
| Molecular Formula | C4H10O |
| Molecular Weight (g/mol) | 74.12 |
| InChI Key | ZXEKIIBDNHEJCQ-UHFFFAOYSA-N |
| IUPAC Name | 2-methylpropan-1-ol |
| SMILES | CC(C)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-335-172
|
Labchem Inc
CF15800015E |
15 gal. |
Each for $9,997.32
|
|
|||||
|
01-335-171
|
Labchem Inc
CF15800020E7 |
20 L |
Each for $4,389.11
|
|
|||||
Specifications
| Colorless | |
| C4H10O | |
| CC(C)CO | |
| 74.12 | |
| Steel Drum | |
| 15 gal. | |
| Liquid |
| 78-83-1,141-97-9 | |
| ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
| 2-methylpropan-1-ol | |
| Reagent | |
| 0.8328 | |
| Non-sterile | |
| Free Lime Reagent |