missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Free Lime Reagent, Reagents
Specifications
Specifications
| Color | Colorless |
| Grade | Reagent |
| Packaging | Steel Drum |
| Specific Gravity | 0.8328 |
| Quantity | 15 gal. |
| Sterility | Non-sterile |
| Physical Form | Liquid |
| Chemical Name or Material | Free Lime Reagent |