missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fosetyl-aluminum

Catalog No. AC467280250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467280250 25 g
AC467281000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467280250 Supplier Thermo Scientific Chemicals Supplier No. 467280250
Only null left

CAS: 39148-24-8 | C6H15AlO9P3, C6H18AlO9P3 | 351.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39148-24-8
Molecular Formula C6H18AlO9P3

Specifications

Quantity 25 g
Solubility Solubility aq. soln.: 120 g/L (20°C). Other solubilities: practically insoluble in acetonitrile, propylene, glycol
Melting Point >300.0°C
Formula Weight 354.11
InChI Key OIPMQULDKWSNGX-UHFFFAOYSA-N
SMILES [Al+3].CCOP(=O)=O.CCOP(=O)=O.CCOP(=O)=O
IUPAC Name aluminium(3+) tris((phosphooxy)ethane)
Molecular Weight (g/mol) 351.08
Chemical Name or Material Fosetyl-Aluminum
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye damage.

Precautionary Statement GHS P Statement Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 254-320-2

RUO – Research Use Only