accessibility menu, dialog, popup

UserName

AlCH3OPOOCH3OPOOCH3OPOO

CAS RN 39148-24-8

IUPAC Name: aluminium(3+) tris((phosphooxy)ethane)
Synonyms: fosetyl-al
Molecular Weight (g/mol): 351.08
Molecular Formula: C6H15AlO9P3
InChi Key: OIPMQULDKWSNGX-UHFFFAOYSA-N
SMILES: [Al+3].CCOP(=O)=O.CCOP(=O)=O.CCOP(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results