missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, FCC, 85%, Spectrum™ Chemical

Catalog No. 18602846
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602846 500 mL Amber Glass Bottle
18602850 2.5 L Glass Bottle
18602847 6 x 500 mL Amber Glass Bottle
18602851 4 x 2.5 L Glass Bottle
18602845 20 L Poly Drum
18602852 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18602846 Supplier Spectrum Chemical Mfg Cor Supplier No. F1135500MLGL
Only null left

F1135, 64-18-6, CH2O2

Spectrum™ Chemical Formic Acid, FCC is the simplest carboxylic acid and one of its biggest uses is as a preservative and antibacterial agent in livestock feed. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
IUPAC Name formic acid
SMILES OC=O

Specifications

CAS Max % 100%
UN Number UN1779
Percent Purity 85%
Grade FCC
Quantity 500 mL
Packaging Amber Glass Bottle