missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 25% v/v, Reagents

Catalog No. 01335180
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335180 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335180 Supplier Labchem Inc Supplier No. CF148100500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formic Acid, 25% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name water
SMILES O

Specifications

pH <2
Boiling Point 100°C
Color Colorless
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.0616
Percent Purity 25%
Grade Reagent
Chemical Name or Material Formic Acid, 25% v/v