Learn More
Mopec™ Formaway Green Liquid Formalin Neutralizer
Catalog No. 22445894
Change view
Click to view available options
Packaging Quantity:
1 gal.
2.5 gal.
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
22445894 5 gal.
22445892 1 gal.
22445893 2.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22445894 Supplier Mopec™ Supplier No. BG5605
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Formaway Green Liquid Formalin Neutralizer quickly and effectively neutralizes spent formalin for sewer disposal. Simply add 32 ounces of Formaway Green to each gallon of 10% neutral buffered formalin waste and allow to stand for at least 4 hours before disposing.
  • Collect formalin waste in the Formaway collection container or equivalent container
  • Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste formalin
  • Add 32 fl. oz. of Formaway Green to each gallon (128 fl. oz.) of 10% NBF formalin waste (For metric users, add 1 liter of Formaway Green to 4 liters of 10% NBF formalin waste).
  • Secure lid on waste collection container and agitate container to mix solutions
  • Allow mixed solutions to stand for approximately 4 hours, adequately reducing the formalin in the waste solution to a non-hazardous safe disposal into the sanitary sewer
  • Repeat step #4, decreasing any potential exposure to the headspace vapor in the collection container
  • Test treated solution with Rapid QC Analysis and carefully pour treated waste into the sanitary sewer
  • Rinse container with cold tap water, preparing it for reuse
Specifications
Material Pre-treated Formalin Neutralizer
Type Formalin Neutralizer
Packaging Quantity 5 gal.