missing translation for 'onlineSavingsMsg'
Learn More

Formamidine disulfide dihydrochloride, 97%

Catalog No. AAB2033122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2033122 100 g
AAB2033106 5 g
AAB2033114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2033122 Supplier Thermo Scientific Chemicals Supplier No. B2033122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14807-75-1 | C2H8Cl2N4S2 | 223.13 g/mol

Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.

Chemical Identifiers

CAS 14807-75-1
Molecular Formula C2H8Cl2N4S2
Molecular Weight (g/mol) 223.13
MDL Number MFCD00012586
InChI Key BFJQSCVWXZOXGK-UHFFFAOYSA-N
Synonym formamidine disulfide dihydrochloride, formamdine disulfide dihydrochloride, dithiourea dihydrochloride, c2h6n4s2.2hcl, carbamimidoyldisulfanylmethanimidamide dihydrochloride, formamidine disulfide dihydrochloride disulfide dihydrochloride
PubChem CID 24802135
IUPAC Name carbamimidoylsulfanyl carbamimidothioate;dihydrochloride
SMILES [Cl-].[Cl-].[NH3+]C(=N)SSC([NH3+])=N

Specifications

Melting Point ∼175°C (decomposition)
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 223.15
Percent Purity 97%
Chemical Name or Material Formamidine disulfide dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-876-3
RTECSNumber LQ4570000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only