missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Buffered, Certified, 10% (v/v), 38 to 42g/L, LabChem™

Supplier:  LabChem LC146704

 View more versions of this product

Catalog No. LC146704


Only null left

Description

Description

Specifications

Specifications

Formaldehyde
7732-18-5 , 50-00-0 , 67-56-1 , 7664-38-2 , 1310-73-2
Phosphorus oxides; Carbon monoxide; Carbon dioxide
Poly Bottle
4 L
30.05
Certified
Buffered
95,3.7,0.5,0.5,0.3
Clear, colorless
HCHO
Formalin
10% (v/v)
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions