missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldéhyde, 37% en poids (histologique), Fisher Chemical
Spécifications
Spécifications
| Nom ou substance chimique | Formaldehyde, 37% by Weight |
| Point de fusion | 0 °C |
| Couleur | Incolore |
| Point d’ébullition | 101 °C |
| Fourchette de pourcentages d’essai | 36,5 à 38,0% |
| Emballage | Bouteille en polyéthylène |
| Formule linéaire | HCHO |
| Quantité | 3,78 L (1 gal) |
| Poids de formule | 30.03 |
| Pourcentage de pureté | 36,5 à 38,0% |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. NFPA Health:3 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |