missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol

CAS: 50-00-0 | CH2O | 30.03 g/mol

$80.60 - $748.62

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA33314AP
View Documents Promotion Details
Thermo Scientific Chemicals
033314AP
500 mL
Each for $80.60
Only null left
 
AA33314K2
View Documents Promotion Details
Thermo Scientific Chemicals
033314K2
1 L
Each for $130.92
Only null left
 
AA33314M1
View Documents Promotion Details
Thermo Scientific Chemicals
033314M1
4 x 1 L
Each for $384.75
Only null left
 
AA33314K7
View Documents Promotion Details
Thermo Scientific Chemicals
033314K7
4 L
Each for $261.14
Only null left
 
AA33314M6
View Documents Promotion Details
Thermo Scientific Chemicals
033314M6
4 x 4 L
Each for $748.62
Only null left
 
Description

Description

Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and is used to make components for transmissions, electrical systems, engine blocks, door panels, axles and brake shoes. It serves as a disinfectant. It is also an ingredient in glues and is used as a preservative in medical laboratories, as an embalming fluid and as a sterilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-00-0
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O

Specifications

50-00-0
1.083 g/mL
60°C (140°F)
CH2O
HCHO
UN1198
14,4235
Soluble in water.
C=O
30.03
CHEBI:16842
36.5 to 38.0%
Liquid
Formaldehyde, 37% in aq. soln., For ACS analysis
°C to 15°C
97°C
36.5-38.0%
1.3765
MFCD00003274
1209228
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
30.03
ACS
Stab. with 10 to 15% methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H370-H351-H314-H226-H317

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H226-H301+H311+H331-H314-H317-H335-H336-H341-H350-H370-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FORMALDEHYDE SOLUTIONS, FLAMMABLE

EINECSNumber : 200-001-8

RTECSNumber : LP8925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only