Learn More
Thermo Scientific Chemicals Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol
CAS: 50-00-0 | CH2O | 30.03 g/mol
Supplier: Thermo Scientific Chemicals 033314K7
| Quantity | 4 L |
|---|
Description
Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and is used to make components for transmissions, electrical systems, engine blocks, door panels, axles and brake shoes. It serves as a disinfectant. It is also an ingredient in glues and is used as a preservative in medical laboratories, as an embalming fluid and as a sterilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| CH2O | |
| MFCD00003274 | |
| formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
| CHEBI:16842 | |
| C=O |
Specifications
| Formaldehyde | |
| 50-00-0 | |
| Colorless | |
| 97°C | |
| Pungent | |
| Stab. with 10 to 15% methanol | |
| 1.3765 | |
| MFCD00003274 | |
| UN1198 | |
| 14,4235 | |
| Soluble in water. | |
| C=O | |
| 30.03 | |
| CHEBI:16842 | |
| 36.5 to 38.0% | |
| Liquid |
| For ACS analysis | |
| −15°C | |
| 1.083 g/mL | |
| 60°C (140°F) | |
| 36.5-38.0% | |
| CH2O | |
| HCHO | |
| 4 L | |
| 1209228 | |
| formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
| WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
| formaldehyde | |
| 712 | |
| 30.03 | |
| ACS |
Safety and Handling
GHS H Statement
H301-H311-H331-H370-H351-H314-H226-H317 Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause an allergic skin reaction.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H226-H301+H311+H331-H314-H317-H335-H336-H341-H350-H370-H372
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FORMALDEHYDE SOLUTIONS, FLAMMABLE
EINECSNumber : 200-001-8
RTECSNumber : LP8925000
TSCA : Yes
Recommended Storage : Ambient temperatures