missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, 37% in aq. soln., ACS, 36.5-38.0%, stab. with 10-15% methanol

Catalog No. AA33314AP
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 x 4 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33314AP 500 mL
AA33314K2 1 L
AA33314M1 4 x 1 L
AA33314K7 4 L
AA33314M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AA33314AP Supplier Thermo Scientific Chemicals Supplier No. 033314AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50-00-0 | CH2O | 30.03 g/mol

Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. It is also used in fiber board, plywood, cigarette smoke, fuel burning appliances, kerosene space heaters and in photography. It acts as a precursor in the manufacturing of automobiles and is used to make components for transmissions, electrical systems, engine blocks, door panels, axles and brake shoes. It serves as a disinfectant. It is also an ingredient in glues and is used as a preservative in medical laboratories, as an embalming fluid and as a sterilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point °C to 15°C
Density 1.083 g/mL
Boiling Point 97°C
Flash Point 60°C (140°F)
Assay Percent Range 36.5-38.0%
Refractive Index 1.3765
Linear Formula HCHO
Quantity 500 mL
UN Number UN1198
Beilstein 1209228
Merck Index 14,4235
Solubility Information Soluble in water.
Formula Weight 30.03
Percent Purity 36.5 to 38.0%
Grade ACS
Physical Form Liquid
Assay Stab. with 10 to 15% methanol
Chemical Name or Material Formaldehyde, 37% in aq. soln., For ACS analysis
Show More Show Less
Hazard Category H226-H301+H311+H331-H314-H317-H335-H336-H341-H350-H370-H372
Hazard Statement GHS H Statement
H301-H311-H331-H370-H351-H314-H226-H317
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FORMALDEHYDE SOLUTIONS, FLAMMABLE
EINECSNumber 200-001-8
RTECSNumber LP8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only