missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 10% (v/v) Solution, Stabilized with approx. 3% Methanol, Histological Grade, Spectrum™ Chemical

Catalog No. p-7165730
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610644 4 L Poly Bottle
18610645 20 L Poly Drum
18610646 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610644 Supplier Spectrum Chemical Mfg Cor Supplier No. HS0084LT
Only null left

Chemical Identifiers

CAS 50-00-0 , 7732-18-5 , 7558-79-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
IUPAC Name formaldehyde
SMILES C=O

Specifications

pH 4.0 to 7.0
Packaging Poly Bottle
Quantity 4 L
Grade Histological

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.