missing translation for 'onlineSavingsMsg'
Learn More

Formalde-Fresh Solution, Buffered, Fisher Chemical™

Catalog No. SF934
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SF934 4 L Poly Bottle
SF9320 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SF934 Supplier Thermo Fisher Scientific Supplier No. SF93 4
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Physical Form Liquid
Chemical Name or Material Formalde-Fresh Solution, Buffered
Boiling Point 100°C
CAS 50-00-0, 67-56-1, 7732-18-5, 7558-79-4
Packaging Poly Bottle
Color Colorless
pH 6.9 to 7.1
Quantity 4 L
MDL Number MFCD00003274
Percent Purity 4.0 to 4.5%