missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Forma™ Freezer Replacement Shelves

Catalog No. 15465394
Change view
Click to view available options
For Use With (Equipment):
13.0 cu. ft. Freezers
17.3 cu. ft. Freezers
Type:
Shelf Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
15465394 17.3 cu. ft. Freezers Shelf Kit
15465393 13.0 cu. ft. Freezers Shelf Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15465394 Supplier Thermo Scientific™ Supplier No. 224135

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.

  • Stainless steel
  • Kits include mounting hardware

Specifications

Type Shelf Kit
Capacity (English) 17.3 cu. ft.
Capacity (Metric) 489.8 L
Style Upright
Color Silver
For Use With (Equipment) 17.3 cu. ft. Freezers