missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Food Yellow No. 4, TCI America™
Description
Food Yellow No. 4 is a chemical compound with the molecular formula C16H9N4Na3O9S2 and a formula weight of 534.36. Key specifications and safety information include:
- Physical Form: Powder
- CAS Registry Number: 1934-21-0
- Acute Toxicity: orl-mus LD50: 12,750 mg/kg; orl-hmn TDLo: 14 μg/kg
- Environmental Toxicity: 48h LC50: >1,000 ppm (Oryzias latipes)
- Safety Hazards: May cause allergy or asthma symptoms or breathing difficulties if inhaled; may cause an allergic skin reaction.
- Storage Conditions: Store in a cool, dark place, kept tightly closed.
- Incompatibilities: Keep away from incompatible materials such as oxidizing agents.
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Physical Form | Powder |
| Formula Weight | 534.36 |
| Chemical Name or Material | Food Yellow No. 4 |
Safety and Handling
| EINECSNumber | (5)-1402 |
| RTECSNumber | UQ6400000 |
| TSCA | Yes |