missing translation for 'onlineSavingsMsg'
Learn More

Food Red No. 3 Aluminum Lake, TCI America™

Catalog No. F017725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F017725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F017725G Supplier TCI America Supplier No. F017725G
Only null left

Chemical Identifiers

CAS 12227-78-0
Molecular Formula C60H18Al2I12O15
Molecular Weight (g/mol) 2555.606
MDL Number MFCD00151206
InChI Key PMKLVGOZGNPKLG-UHFFFAOYSA-H
PubChem CID 24832094
IUPAC Name dialuminum;2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C(=C4OC5=C(C(=C(C=C35)I)[O-])I)I)[O-])I.C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C(=C4OC5=C(C(=C(C=C35)I)[O-])I)I)[O-])I.C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C(=C4OC5=C(C(=C(C=C35)I)[O-])I)I)[O-])I.[Al+3].[Al+3]

Specifications

Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Food Red No. 3 Aluminum Lake
EINECSNumber (5)-3274
TSCA Yes