missing translation for 'onlineSavingsMsg'
Learn More

Folic Acid Impurity F Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002338
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002338 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002338 Supplier Merck Emd Millipore Supplier No. PHR200320MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 1391194-56-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C7H6 ClN5 O
Synonym 2-Amino-7-(chloromethyl)pteridin-4(1 H)-one
Type Certified Reference Material
Quantity 20 mg
Formula Weight 211.61
Grade Certified Reference Material