missing translation for 'onlineSavingsMsg'
Learn More

FMOC-O-tert-Butyl-L-tyrosine, 98%

Numéro de catalogue. AC439090100
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC439090100 10 g
AC439090250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC439090100 Fournisseur Thermo Scientific Chemicals Code fournisseur 439090100
Il en reste null

CAS: 71989-38-3 | C28H29NO5 | 459.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 71989-38-3
Molecular Formula C28H29NO5
Molecular Weight (g/mol) 459.54
MDL Number MFCD00037129
InChI Key JAUKCFULLJFBFN-KSYWNVGFNA-N
Synonym fmoc-tyr tbu-oh, fmoc-o-tert-butyl-l-tyrosine, fmoc-tyr but-oh, fmoc-tyr t-bu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, fmoc-tyr but, n-fmoc-o-tert-butyl-l-tyrosine, nalpha-fmoc-o-tert-butyl-l-tyrosine, n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine, 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 10895791
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
SMILES CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

Spécifications

Melting Point 150°C to 156°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 10 g
Specific Rotation -29.5
Formula Weight 459.55
Percent Purity 98%
Physical Form Powder
Chemical Name or Material FMOC-O-tert-Butyl-L-tyrosine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 276-262-7

RUO – Research Use Only