missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FMO3 Recombinant Rabbit Monoclonal Antibody (6Q10X9)

Description
Positive Samples: Mouse lung, Mouse liver, Mouse kidney, Rat liver, Rat kidney Immunogen sequence: KPNVKEFTET SAIFEDGTIF EGIDCVIFAT GYSFAYPFLD ESIIKSRNNE IILFKGVFPP LLEKSTIAVI GFVQSLGAAI PTVDLQSRWA AQVIKGTCTL P.
The mammalian flavin-containing monooxygenases (FMO; EC 1.14.13.8) represent a multigene family whose gene products are localized in the endoplasmic reticulum of many tissues. These enzymes catalyze the NADPH-dependent oxidative metabolism of many drugs, pesticides, and other foreign compounds. Their substrates are soft nucleophiles with an electron-rich center, typically a nitrogen, sulfur or phosphorus-containing functional group, as the site for oxidative attack by the enzyme.
Spécifications
Spécifications
| Antigen | FMO3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 6Q10X9 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Fmo3 |
| Gene Accession No. | P31513, P97501, Q9EQ76 |
| Gene Alias | AW111792; Dimethylaniline monooxygenase [N-oxide-forming] 3; dimethylaniline oxidase 3; dJ127D3.1; flavin containing dimethylaniline monoxygenase 3; flavin containing monooxygenase 3; flavin-containing monooxygenase 3; flavin-containing monooxygenase FMO3; FM03; FMO 1D1; FMO 3; FMO form 2; FMO II; FMO3; FMOII; hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; MGC34400; TMAU; Trimethylamine monooxygenase; Unknown (protein for MGC:128044) |
| Afficher plus de résultats |